Assay Design
Gene Transcripts
- Gene Symbol
- Zfp58
- Entrez Gene ID
-
238693
- Gene Name
- zinc finger protein 58
- Gene Aliases
-
A530094I17Rik, Mfg-1, Mfg1, Rslcan5, Zfp817
- Chromosome Location
-
Chr.13: 67490167 - 67500522 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK167399.1 |
- |
1 - 2 |
100 |
96 |
View Full Product Details