Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138676.2 | NP_619617.2 | 11 - 12 | 1103 | 82 |
GenBank mRNA | AF246218.1 | - | 11 - 12 | 1109 | 82 |
AK047434.1 | - | 9 - 10 | 1234 | 82 | |
AK154779.1 | - | 11 - 12 | 1091 | 82 | |
AK160641.1 | - | 10 - 11 | 1175 | 82 | |
AK198381.1 | - | 1 - 2 | 97 | 82 | |
AK203091.1 | - | 1 - 2 | 53 | 82 | |
BC061477.1 | - | 11 - 12 | 1056 | 82 |