Assay Design



Gene Transcripts

Gene Symbol
Palm
Entrez Gene ID
18483
Gene Name
paralemmin
Gene Aliases
-
Chromosome Location
Chr.10: 79793351 - 79820896 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023128.4 NP_075617.3 6 - 7 608 98
GenBank mRNA AK037044.1 - 6 - 7 465 98
AK129107.1 - 4 - 5 347 98
Y14771.1 - 6 - 7 571 98

View Full Product Details