Assay Design
Gene Transcripts
- Gene Symbol
- Tbata
- Entrez Gene ID
-
65971
- Gene Name
- thymus, brain and testes associated
- Gene Aliases
-
1700021K02Rik, AI428928, Spatial, Titest
- Chromosome Location
-
Chr.10: 61171964 - 61188841 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK006213.1 |
- |
4 - 5 |
460 |
133 |
View Full Product Details