Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017419.2 | NP_001017419.1 | 6 - 7 | 903 | 125 |
NM_001017433.2 | NP_001017433.1 | 6 - 7 | 1005 | 125 | |
GenBank mRNA | AF521591.1 | - | 5 - 6 | 834 | 125 |
AF521592.1 | - | 5 - 6 | 936 | 125 |