Assay Design



Gene Transcripts

Gene Symbol
Prss57
Entrez Gene ID
73106
Gene Name
protease, serine 57
Gene Aliases
2900092M14Rik, GLGL782, Prssl1, UNQ782
Chromosome Location
Chr.10: 79781474 - 79795605 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK013841.1 - 1 - 2 221 110

View Full Product Details