Assay Design



Gene Transcripts

Gene Symbol
Micu1
Entrez Gene ID
216001
Gene Name
mitochondrial calcium uptake 1
Gene Aliases
C730016L05Rik, Calc, Cbara1
Chromosome Location
Chr.10: 59702482 - 59864134 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291442.1 NP_001278371.1 3 - 4 465 67
NM_001291443.1 NP_001278372.1 3 - 4 465 67
NM_144822.3 NP_659071.1 3 - 4 465 67
GenBank mRNA AK046049.1 - 3 - 4 430 67
AK049521.1 - 3 - 4 461 67
AK050113.1 - 3 - 4 452 67
AK051470.1 - 3 - 4 371 67
AK052485.1 - 3 - 4 413 67
AK077605.1 - 3 - 4 415 67
AK083520.1 - 3 - 4 468 67
BC018320.1 - 3 - 4 406 67
BC023022.1 - 3 - 4 400 67
BC023042.1 - 2 - 3 349 67
BC026566.1 - 3 - 4 416 67

View Full Product Details