Assay Design



Gene Transcripts

Gene Symbol
Rora
Entrez Gene ID
19883
Gene Name
RAR-related orphan receptor alpha
Gene Aliases
9530021D13Rik, Nr1f1, ROR1, ROR2, ROR3, nmf267, sg, staggerer, tmgc26
Chromosome Location
Chr.9: 68653402 - 69388246 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013646.2 NP_038674.1 1 - 2 187 64
GenBank mRNA U53228.1 - 1 - 2 171 64

View Full Product Details