Assay Design



Gene Transcripts

Gene Symbol
Yap1
Entrez Gene ID
22601
Gene Name
yes-associated protein 1
Gene Aliases
AI325207, Yap, Yap65, Yki, Yorkie
Chromosome Location
Chr.9: 7931999 - 8004596 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171147.1 NP_001164618.1 6 - 7 1188 72
GenBank mRNA BC014733.1 - 6 - 7 1185 72

View Full Product Details