Assay Design



Gene Transcripts

Gene Symbol
Aes
Entrez Gene ID
14797
Gene Name
amino-terminal enhancer of split
Gene Aliases
AL024115, Esp1, Grg, Grg-5, Grg5, Tle5
Chromosome Location
Chr.10: 81559444 - 81566371 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276288.1 NP_001263217.1 6 - 7 566 139
GenBank mRNA BC145299.1 - 6 - 7 454 139

View Full Product Details