Assay Design
Gene Transcripts
- Gene Symbol
- Sumo3
- Entrez Gene ID
-
20610
- Gene Name
- SMT3 suppressor of mif two 3 homolog 3 (yeast)
- Gene Aliases
-
2810014B19Rik, D10Ertd345e, SMT3A, SUMO-3, Smt3h1
- Chromosome Location
-
Chr.10: 77606097 - 77618331 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_125904.1 |
|
4 - 5 |
555 |
123 |
GenBank mRNA |
CK619326.1 |
- |
4 - 5 |
415 |
123 |
View Full Product Details