Assay Design
Gene Transcripts
- Gene Symbol
- Umps
- Entrez Gene ID
-
22247
- Gene Name
- uridine monophosphate synthetase
- Gene Aliases
-
1700095D23Rik, AA408257, AL033308, BB164745
- Chromosome Location
-
Chr.16: 33954784 - 33967302 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK035997.1 |
- |
1 - 2 |
181 |
60 |
View Full Product Details