Assay Design



Gene Transcripts

Gene Symbol
Gnb1l
Entrez Gene ID
13972
Gene Name
guanine nucleotide binding protein (G protein), beta polypeptide 1-like
Gene Aliases
ESTM55, Gm16314, Me49f07, OTTMUSG00000033458, Wdr14, Wdvcf
Chromosome Location
Chr.16: 18498021 - 18566680 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285494.1 NP_001272423.1 6 - 7 703 62
GenBank mRNA AK166195.1 - 6 - 7 687 62

View Full Product Details