Assay Design
Gene Transcripts
- Gene Symbol
- Slc51b
- Entrez Gene ID
-
330962
- Gene Name
- solute carrier family 51, beta subunit
- Gene Aliases
-
Ostb
- Chromosome Location
-
Chr.9: 65412753 - 65422773 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK002463.1 |
- |
1 - 2 |
161 |
72 |
AK004188.1 |
- |
1 - 2 |
213 |
72 |
View Full Product Details