Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178933.2 | NP_849264.1 | 3 - 4 | 239 | 90 |
GenBank mRNA | AK002463.1 | - | 3 - 4 | 370 | 90 |
AK004188.1 | - | 3 - 4 | 422 | 90 | |
AY279396.1 | - | 3 - 4 | 239 | 90 | |
BC115606.1 | - | 2 - 3 | 218 | 90 | |
BC115607.1 | - | 2 - 3 | 218 | 90 |