Assay Design



Gene Transcripts

Gene Symbol
Parp16
Entrez Gene ID
214424
Gene Name
poly (ADP-ribose) polymerase family, member 16
Gene Aliases
ARTD15, BC055447, C79952, PARP-16
Chromosome Location
Chr.9: 65214380 - 65239220 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177460.4 NP_803411.3 1 - 2 177 65
GenBank mRNA AK133475.1 - 1 - 2 178 65
AK169866.1 - 1 - 2 92 65
BC055447.1 - 1 - 2 146 65

View Full Product Details