Assay Design



Gene Transcripts

Gene Symbol
Fbxo22
Entrez Gene ID
71999
Gene Name
F-box protein 22
Gene Aliases
0610033L19Rik, 1600016C16Rik
Chromosome Location
Chr.9: 55208935 - 55224433 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028049.2 NP_082325.2 3 - 4 483 72
GenBank mRNA AK002745.1 - 3 - 4 363 72
AK005473.1 - 2 - 3 311 72
AK181573.1 - 2 - 3 143 72
AK184767.1 - 2 - 3 143 72
BC018273.1 - 3 - 4 423 72

View Full Product Details