Assay Design



Gene Transcripts

Gene Symbol
Fbxo22
Entrez Gene ID
71999
Gene Name
F-box protein 22
Gene Aliases
0610033L19Rik, 1600016C16Rik
Chromosome Location
Chr.9: 55208935 - 55224433 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028049.2 NP_082325.2 6 - 7 906 70
GenBank mRNA AK002745.1 - 6 - 7 786 70
AK005473.1 - 5 - 6 734 70
AK207864.1 - 2 - 3 344 70
AK211102.1 - 2 - 3 208 70
AK213133.1 - 2 - 3 341 70
AK219248.1 - 2 - 3 341 70
BC018273.1 - 6 - 7 846 70

View Full Product Details