Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027912.1 | NP_082188.1 | 2 - 3 | 1361 | 65 |
GenBank mRNA | AK005069.1 | - | 2 - 3 | 1362 | 65 |
AK154513.1 | - | 2 - 3 | 1344 | 65 | |
BC119320.1 | - | 2 - 3 | 1339 | 65 |