Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144933.1 | NP_659182.1 | 1 - 2 | 317 | 145 |
GenBank mRNA | AK033151.1 | - | 1 - 2 | 323 | 145 |
AK038451.1 | - | 1 - 2 | 357 | 145 | |
AK049112.1 | - | 1 - 2 | 357 | 145 | |
AK142033.1 | - | 1 - 2 | 356 | 146 | |
AK151377.1 | - | 1 - 2 | 357 | 145 | |
BC020185.1 | - | 1 - 2 | 317 | 145 |