Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011734.3 | NP_035864.2 | 7 - 8 | 1058 | 100 |
GenBank mRNA | AF156856.1 | - | 8 - 9 | 1260 | 100 |
AK028656.1 | - | 7 - 8 | 1058 | 100 | |
AK033980.1 | - | - | 306 | 100 | |
AK035715.1 | - | 5 - 6 | 792 | 100 | |
AK158634.1 | - | 6 - 7 | 996 | 100 | |
AK164852.1 | - | 7 - 8 | 1065 | 100 | |
AK165103.1 | - | 7 - 8 | 1085 | 100 | |
AK167103.1 | - | 7 - 8 | 1074 | 100 | |
BC007136.1 | - | 7 - 8 | 1076 | 100 | |
U40408.1 | - | 7 - 8 | 1064 | 100 | |
U61183.1 | - | 7 - 8 | 1057 | 100 | |
X98625.1 | - | 7 - 8 | 1057 | 100 |