Assay Design



Gene Transcripts

Gene Symbol
Clip3
Entrez Gene ID
76686
Gene Name
CAP-GLY domain containing linker protein 3
Gene Aliases
1500005P14Rik, AI844915, Clipr-59
Chromosome Location
Chr.7: 30291688 - 30308367 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081114.1 NP_001074583.1 12 - 13 1667 77
GenBank mRNA AK005167.1 - 4 - 5 464 77
BC056173.1 - 4 - 5 435 77
BC138413.1 - 11 - 12 1586 77

View Full Product Details