Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007691.5 | NP_031717.2 | 3 - 4 | 407 | 85 |
GenBank mRNA | AF016583.1 | - | 2 - 3 | 316 | 85 |
AK011258.1 | - | 3 - 4 | 351 | 85 | |
AK033179.1 | - | 3 - 4 | 377 | 85 | |
AK045336.1 | - | 3 - 4 | 349 | 85 | |
AK166675.1 | - | 3 - 4 | 408 | 85 | |
BC053394.1 | - | 3 - 4 | 343 | 85 | |
BC064445.1 | - | 2 - 3 | 807 | 85 | |
BC100386.1 | - | 2 - 3 | 482 | 85 |