Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007691.5 | NP_031717.2 | 9 - 10 | 1040 | 121 |
GenBank mRNA | AF016583.1 | - | 8 - 9 | 949 | 121 |
AF032875.1 | - | 3 - 4 | 318 | 121 | |
AK011258.1 | - | 9 - 10 | 984 | 121 | |
AK033179.1 | - | 9 - 10 | 1010 | 121 | |
AK045336.1 | - | 9 - 10 | 982 | 121 | |
AK166675.1 | - | 9 - 10 | 1041 | 121 | |
BC037613.1 | - | 8 - 9 | 753 | 121 | |
BC053394.1 | - | 9 - 10 | 977 | 121 | |
BC064445.1 | - | 8 - 9 | 1439 | 121 | |
BC100386.1 | - | 8 - 9 | 1115 | 121 |