Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178374.3 | NP_848461.1 | 6 - 7 | 644 | 138 |
GenBank mRNA | AK002717.1 | - | 6 - 7 | 660 | 138 |
AK013151.1 | - | 6 - 7 | 654 | 138 | |
AK155315.1 | - | 6 - 7 | 661 | 138 | |
BC023317.1 | - | - | 624 | 138 | |
BC049638.1 | - | 6 - 7 | 644 | 138 | |
BC061258.1 | - | 6 - 7 | 649 | 138 |