Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019651.2 | NP_062625.2 | 11 - 12 | 1989 | 101 |
GenBank mRNA | AF013490.1 | - | 11 - 12 | 1459 | 101 |
AK089069.1 | - | 11 - 12 | 1563 | 101 | |
BC053017.1 | - | 11 - 12 | 1989 | 101 |