Assay Design



Gene Transcripts

Gene Symbol
Ptpn9
Entrez Gene ID
56294
Gene Name
protein tyrosine phosphatase, non-receptor type 9
Gene Aliases
MEG2
Chromosome Location
Chr.9: 56994968 - 57062808 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019651.2 NP_062625.2 9 - 10 1755 74
GenBank mRNA AK089069.1 - 9 - 10 1329 74
BC053017.1 - 9 - 10 1755 74

View Full Product Details