Assay Design



Gene Transcripts

Gene Symbol
Fxyd2
Entrez Gene ID
11936
Gene Name
FXYD domain-containing ion transport regulator 2
Gene Aliases
Atp1g1
Chromosome Location
Chr.9: 45399709 - 45410278 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007503.3 NP_031529.2 2 - 3 127 138
GenBank mRNA AK002215.1 - 2 - 3 115 138
AK002228.1 - 2 - 3 128 138
AY626243.1 - 2 - 3 249 138
AY626245.1 - 2 - 3 285 174
BC024614.1 - 2 - 3 116 138
BU756853.1 - 4 - 5 138

View Full Product Details