Assay Design



Gene Transcripts

Gene Symbol
Rdx
Entrez Gene ID
19684
Gene Name
radixin
Gene Aliases
AA516625
Chromosome Location
Chr.9: 52047150 - 52088738 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104617.1 NP_001098087.1 2 - 3 563 128
NM_009041.3 NP_033067.2 2 - 3 563 128
GenBank mRNA AK016711.1 - 2 - 3 234 128
AK088292.1 - 2 - 3 270 128
AK146423.1 - 2 - 3 221 128
AK156370.1 - 2 - 3 249 128
AK168449.1 - 2 - 3 234 128
AK217469.1 - 2 - 3 101 128
BC002041.1 - 2 - 3 208 128
BC026786.1 - 2 - 3 236 128
BC034438.1 - 2 - 3 389 128
BC053417.1 - 2 - 3 237 128
X60672.1 - 2 - 3 176 128

View Full Product Details