Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281954.1 | NP_001268883.1 | 2 - 3 | 228 | 75 |
NM_019465.4 | NP_062338.3 | 2 - 3 | 228 | 75 | |
GenBank mRNA | AF001104.1 | - | 2 - 3 | 186 | 75 |
BC117715.1 | - | 2 - 3 | 186 | 75 | |
BC117716.1 | - | 2 - 3 | 186 | 75 |