Assay Design



Gene Transcripts

Gene Symbol
Prtg
Entrez Gene ID
235472
Gene Name
protogenin homolog (Gallus gallus)
Gene Aliases
A230098A12Rik, Igdcc5
Chromosome Location
Chr.9: 72807274 - 72917307 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175485.4 NP_780694.3 16 - 17 2973 76
GenBank mRNA AK039115.1 - 3 - 4 489 76
AK083540.1 - 14 - 15 2409 76
AY630257.1 - 16 - 17 3043 76
BC116336.1 - 14 - 15 2406 76
BC116337.1 - 14 - 15 2406 76
DQ360114.1 - 16 - 17 2973 76

View Full Product Details