Assay Design



Gene Transcripts

Gene Symbol
Prtg
Entrez Gene ID
235472
Gene Name
protogenin homolog (Gallus gallus)
Gene Aliases
A230098A12Rik, Igdcc5
Chromosome Location
Chr.9: 72807274 - 72917307 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175485.4 NP_780694.3 2 - 3 561 123
GenBank mRNA AY630257.1 - 2 - 3 631 123
DQ360114.1 - 2 - 3 561 123

View Full Product Details