Assay Design



Gene Transcripts

Gene Symbol
Zglp1
Entrez Gene ID
100009600
Gene Name
zinc finger, GATA-like protein 1
Gene Aliases
Glp1
Chromosome Location
Chr.9: 21062393 - 21067093 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103168.1 NP_001096638.1 3 - 4 588 68
GenBank mRNA DQ286956.1 - 3 - 4 588 68

View Full Product Details