Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190911.1 | NP_001177840.1 | 11 - 12 | 1727 | 83 |
NM_001190912.1 | NP_001177841.1 | 10 - 11 | 1649 | 83 | |
NM_001190913.1 | NP_001177842.1 | 11 - 12 | 1727 | 83 | |
NM_001190914.1 | NP_001177843.1 | 12 - 13 | 1828 | 83 | |
NM_026324.3 | NP_080600.1 | 11 - 12 | 1727 | 83 | |
GenBank mRNA | AK034142.1 | - | 11 - 12 | 1664 | 83 |
AK038385.1 | - | 11 - 12 | 1478 | 83 | |
AK043580.1 | - | 2 - 3 | 200 | 83 | |
AK045373.1 | - | 11 - 12 | 1632 | 83 | |
AK079332.1 | - | 12 - 13 | 1766 | 83 | |
AK171866.1 | - | 8 - 9 | 1078 | 83 | |
AY169782.1 | - | 11 - 12 | 1727 | 83 | |
BC063072.1 | - | 10 - 11 | 1444 | 83 |