Assay Design



Gene Transcripts

Gene Symbol
Kirrel3
Entrez Gene ID
67703
Gene Name
kin of IRRE like 3 (Drosophila)
Gene Aliases
1500010O20Rik, 2900036G11Rik, NEPH2, SST4, mKIAA1867
Chromosome Location
Chr.9: 34485852 - 35036315 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190911.1 NP_001177840.1 15 - 16 2181 61
NM_001190912.1 NP_001177841.1 14 - 15 2178 61
NM_001190914.1 NP_001177843.1 15 - 16 2246 61
NM_026324.3 NP_080600.1 14 - 15 2145 61
GenBank mRNA AK045373.1 - 15 - 16 2086 61
AK079332.1 - 15 - 16 2184 61
AK171866.1 - 11 - 12 1496 61
AY169782.1 - 14 - 15 2145 61
BC063072.1 - 14 - 15 1973 61

View Full Product Details