Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190911.1 | NP_001177840.1 | 10 - 11 | 1622 | 112 |
NM_001190912.1 | NP_001177841.1 | 9 - 10 | 1544 | 112 | |
NM_001190913.1 | NP_001177842.1 | 10 - 11 | 1622 | 112 | |
NM_001190914.1 | NP_001177843.1 | 11 - 12 | 1723 | 112 | |
NM_026324.3 | NP_080600.1 | 10 - 11 | 1622 | 112 | |
GenBank mRNA | AK034142.1 | - | 10 - 11 | 1559 | 112 |
AK038385.1 | - | 10 - 11 | 1373 | 112 | |
AK043580.1 | - | 1 - 2 | 95 | 112 | |
AK045373.1 | - | 10 - 11 | 1527 | 112 | |
AK079332.1 | - | 11 - 12 | 1661 | 112 | |
AK171866.1 | - | 7 - 8 | 973 | 112 | |
AY169782.1 | - | 10 - 11 | 1622 | 112 | |
BC063072.1 | - | 9 - 10 | 1339 | 112 |