Assay Design
Gene Transcripts
- Gene Symbol
- Ift46
- Entrez Gene ID
-
76568
- Gene Name
- intraflagellar transport 46
- Gene Aliases
-
1500035H01Rik, AA408110, AI663846, R75047, c11orf2
- Chromosome Location
-
Chr.9: 44772938 - 44792714 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC010326.1 |
- |
2 - 3 |
145 |
75 |
BC080764.1 |
- |
3 - 4 |
288 |
75 |
View Full Product Details