Assay Design



Gene Transcripts

Gene Symbol
Slc16a9
Entrez Gene ID
66859
Gene Name
solute carrier family 16 (monocarboxylic acid transporters), member 9
Gene Aliases
1200003C15Rik, 4930425B13Rik, AI552636
Chromosome Location
Chr.10: 70245276 - 70285951 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025807.3 NP_080083.2 1 - 2 100 98
GenBank mRNA AK004556.1 - 1 - 2 101 98
AK004599.1 - 1 - 2 279 98
AK004684.2 - 1 - 2 158 98

View Full Product Details