Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162475.1 | NP_001155947.1 | 18 - 19 | 1921 | 55 |
NM_001162476.1 | NP_001155948.1 | 18 - 19 | 2232 | 55 | |
NM_001162479.1 | NP_001155951.1 | 18 - 19 | 2026 | 55 | |
NM_009291.2 | NP_033317.2 | 18 - 19 | 2191 | 55 | |
GenBank mRNA | AF062476.1 | - | 18 - 19 | 2191 | 55 |
BC075657.1 | - | 18 - 19 | 1921 | 55 |