Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133883.2 | NP_598644.1 | 2 - 3 | 358 | 78 |
GenBank mRNA | AJ427296.1 | - | 2 - 3 | 358 | 78 |
AK143583.1 | - | 2 - 3 | 348 | 78 | |
AK147034.1 | - | 2 - 3 | 334 | 78 | |
AK149904.1 | - | 2 - 3 | 350 | 78 | |
AK150202.1 | - | 2 - 3 | 350 | 78 | |
AK169070.1 | - | 2 - 3 | 345 | 78 | |
AK171780.1 | - | 2 - 3 | 348 | 78 | |
BC023177.1 | - | 2 - 3 | 335 | 78 | |
BC028965.1 | - | 2 - 3 | 122 | 78 | |
BC045203.1 | - | 2 - 3 | 396 | 78 |