Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172701.2 | NP_766289.2 | 7 - 8 | 934 | 76 |
GenBank mRNA | AB154826.1 | - | 7 - 8 | 887 | 76 |
AB286960.1 | - | 8 - 9 | 935 | 76 | |
AK077467.1 | - | 7 - 8 | 900 | 76 | |
AK077909.1 | - | 7 - 8 | 855 | 76 | |
BC045150.1 | - | 7 - 8 | 880 | 76 |