Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023603.3 | NP_076092.1 | 9 - 10 | 71 | |
GenBank mRNA | AK018483.1 | - | 4 - 5 | 71 | |
AK191209.1 | - | 2 - 3 | 71 | ||
AK198254.1 | - | 2 - 3 | 71 | ||
AK217233.1 | - | 3 - 4 | 71 | ||
AK219185.1 | - | 2 - 3 | 71 | ||
AY034062.1 | - | 9 - 10 | 71 | ||
BC037486.1 | - | 7 - 8 | 71 | ||
BC049227.1 | - | 6 - 7 | 71 | ||
BC055468.1 | - | 5 - 6 | 71 | ||
BC080848.1 | - | 9 - 10 | 2041 | 69 | |
BC089305.1 | - | 9 - 10 | 71 |