Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023665.3 | NP_076154.3 | 3 - 4 | 886 | 144 |
GenBank mRNA | AK012363.1 | - | 3 - 4 | 887 | 144 |
AK150024.1 | - | 3 - 4 | 887 | 144 | |
AK150665.1 | - | 3 - 4 | 866 | 144 | |
AK152736.1 | - | 3 - 4 | 866 | 144 | |
AK178549.1 | - | 2 - 3 | 232 | 144 | |
AK187384.1 | - | 2 - 3 | 145 |