Assay Design



Gene Transcripts

Gene Symbol
Sptbn1
Entrez Gene ID
20742
Gene Name
spectrin beta, non-erythrocytic 1
Gene Aliases
9930031C03Rik, AL033301, SPTB2, Spnb-2, Spnb2, elf1, elf3, mKIAA4049
Chromosome Location
Chr.11: 30099395 - 30268204 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009260.2 NP_033286.2 19 - 20 4657 85
NM_175836.2 NP_787030.2 19 - 20 4317 85
GenBank mRNA AF017112.1 - 19 - 20 4564 85
AK220456.1 - 20 - 21 4519 85
U73171.1 - 1 - 2 97 85

View Full Product Details