Assay Design



Gene Transcripts

Gene Symbol
Sptbn1
Entrez Gene ID
20742
Gene Name
spectrin beta, non-erythrocytic 1
Gene Aliases
9930031C03Rik, AL033301, SPTB2, Spnb-2, Spnb2, elf1, elf3, mKIAA4049
Chromosome Location
Chr.11: 30099395 - 30268204 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009260.2 NP_033286.2 24 - 25 5584 71
NM_175836.2 NP_787030.2 24 - 25 5244 71
GenBank mRNA AF017112.1 - 24 - 25 5491 71
AK161119.1 - 3 - 4 367 71
AK220456.1 - 25 - 26 5446 71
M74773.1 - 24 - 25 5564 71

View Full Product Details