Assay Design



Gene Transcripts

Gene Symbol
Ctps
Entrez Gene ID
51797
Gene Name
cytidine 5'-triphosphate synthase
Gene Aliases
Ctps1
Chromosome Location
Chr.4: 120539866 - 120570276 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016748.2 NP_058028.2 1 - 2 128 106
GenBank mRNA AK146476.1 - 1 - 2 129 106
AK148259.1 - 1 - 2 77 106
AK161110.1 - 1 - 2 76 106
AK164232.1 - 1 - 2 118 106
BC006698.1 - 1 - 2 103 106

View Full Product Details