Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146153.3 | NP_666265.3 | 10 - 11 | 2792 | 76 |
GenBank mRNA | AK046010.1 | - | 2 - 3 | 1125 | 76 |
AK153954.1 | - | 4 - 5 | 591 | 76 | |
AK169008.1 | - | 4 - 5 | 231 | 76 | |
BC033472.1 | - | 2 - 3 | 389 | 76 | |
BC040346.1 | - | 10 - 11 | 2723 | 76 | |
BC048391.1 | - | 3 - 4 | 158 | 76 | |
BC092239.1 | - | 10 - 11 | 2708 | 76 |