Assay Design
Gene Transcripts
- Gene Symbol
- Hormad2
- Entrez Gene ID
-
75828
- Gene Name
- HORMA domain containing 2
- Gene Aliases
-
4930529M09Rik
- Chromosome Location
-
Chr.11: 4345801 - 4442459 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK015939.1 |
- |
3 - 4 |
319 |
80 |
View Full Product Details