Assay Design



Gene Transcripts

Gene Symbol
Slc22a16
Entrez Gene ID
70840
Gene Name
solute carrier family 22 (organic cation transporter), member 16
Gene Aliases
4921504E14Rik, CT2, FLIPT2, OCT6, OKB1
Chromosome Location
Chr.10: 40570321 - 40604135 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027572.1 NP_081848.1 4 - 5 1291 86
GenBank mRNA AK014809.1 - 1 - 2 524 86
BC100473.1 - 4 - 5 1291 86
BC138326.1 - 4 - 5 1302 86
BC138327.1 - 4 - 5 1302 86
DQ164804.2 - 4 - 5 1317 86
DQ164805.1 - 4 - 5 1278 86

View Full Product Details