Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001495.2 | NP_001001495.2 | 4 - 5 | 490 | 123 |
GenBank mRNA | AK033534.1 | - | 4 - 5 | 294 | 123 |
BC104392.1 | - | 4 - 5 | 293 | 123 | |
BC104393.1 | - | 4 - 5 | 293 | 123 | |
EF585494.1 | - | 4 - 5 | 472 | 123 | |
EF585495.1 | - | 4 - 5 | 472 | 123 |